2,3-Dimethoxyzimtsäure
Lieferant: MENAI
Synonyme:
2,3-Dimethoxycinnamsäure, 3-(2,3-Dimethoxyphenyl)propensäure
DH586.100
MENADH586.100EA
0
EUR
MENADH586.100
2,3-Dimethoxyzimtsäure
2,3-Dimethoxyzimtsäure
Formel:
C₁₁H₁₂O₄ Molecular Weight: 208.21 g/mol |
CAS-Nummer:
7461-60-1 EINECS: 231-249-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...