2,4-Dichlorbenzhydrazid

Lieferant: MENAI

Synonyme: 2,4-Dichlorobenzoic hydrazide

DH604.25
MENADH604.25EA 0 EUR
MENADH604.25
2,4-Dichlorbenzhydrazid
2,4-Dichlorbenzhydrazid
Formel: C₇H₆Cl₂N₂O
Molecular Weight: 205,04 g/mol
MDL: MFCD00025113
CAS-Nummer: 5814-06-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR