5-Chlor-7-methylisatin

Lieferant: MENAI

Synonyme: 5-Chlor-7-methylindolin-2,3-dion, 5-Chlor-7-methyl-1H-indol-2,3-dion, 5-Chlor-7-methyl-2,3-indolindion, 5-Chlor-7-methyl-2,3-dioxoindolin

DH612.5
MENADH612.5EA 0 EUR
MENADH612.5
5-Chlor-7-methylisatin
5-Chlor-7-methylisatin
Formel: C₉H₆ClNO₂
Molecular Weight: 195,6 g/mol
MDL: MFCD00047218
CAS-Nummer: 14389-06-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR