5-(Trifluormethyl)-o-anisidin
Lieferant: MENAI
Synonyme:
3-Amino-4-methoxybenzotrifluorid, 5-(Trifluormethyl)-2-methoxyanilin, 2-Methoxy-5-(trifluormethyl)anilin, 2-Amino-4-(trifluormethyl)anisol, 5-(Trifluormethyl)-2-anisidin
DH624.10
MENADH624.10EA
0
EUR
MENADH624.10
5-(Trifluormethyl)-o-anisidin
5-(Trifluormethyl)-o-anisidin
Formel:
C₈H₈F₃NO Molecular Weight: 191,15 g/mol Siedepunkt: 230 °C (1013 hPa) Schmelzpunkt: 59 °C |
MDL:
MFCD00042486 CAS-Nummer: 349-65-5 UN: 2811 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...