4-(1-Piperidinyl)anilin

Lieferant: MENAI

Synonyme: 4-Piperidinoaniline

DH625.10
MENADH625.10EA 0 EUR
MENADH625.10
4-(1-Piperidinyl)anilin
4-(1-Piperidinyl)anilin
Formel: C₁₁H₁₆N₂
Molecular Weight: 176,26 g/mol
MDL: MFCD00051688
CAS-Nummer: 2359-60-6
UN: 2811
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR