3-Phenoxybenzylchlorid
Lieferant: MENAI
Synonyme:
1-Chlormethyl-3-phenoxybenzol
E1051.25
MENAE1051.25EA
0
EUR
MENAE1051.25
3-Phenoxybenzylchlorid
3-Phenoxybenzylchlorid
Formel:
C₁₃H₁₁ClO Molecular Weight: 218,68 g/mol Siedepunkt: 123 °C (0,3 mmHg) Dichte: 1,16 g/cm³ (20 °C) |
MDL:
MFCD00040866 CAS-Nummer: 53874-66-1 EINECS: 258-831-1 UN: 3265 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...