3-(Trifluormethoxy)zimtsäure

Lieferant: MENAI

Synonyme: 3-(3-(Trifluoromethoxy)phenyl)acrylic acid

E1781.10
MENAE1781.10EA 0 EUR
MENAE1781.10
3-(Trifluormethoxy)zimtsäure
3-(Trifluormethoxy)zimtsäure
Formel: C₁₀H₇F₃O₃
Molecular Weight: 232,16 g/mol
MDL: MFCD00066337
CAS-Nummer: 168833-80-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR