2-Chlor-6-fluorzimtsäure

Lieferant: MENAI

E1843.100 E1843.50
MENAE1843.100EA 0 EUR
MENAE1843.100 MENAE1843.50
2-Chlor-6-fluorzimtsäure
2-Chlor-6-fluorzimtsäure
Formel: C₉H₆ClFO₂
MDL: MFCD00051582
CAS-Nummer: 392-22-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR