2-Pentylthiophen

Lieferant: MENAI

E2525.10
MENAE2525.10EA 0 EUR
MENAE2525.10
2-Pentylthiophen
2-Pentylthiophen
Formel: C₉H₁₄S
Molecular Weight: 154.28 g/mol
MDL: MFCD00041017
CAS-Nummer: 4861-58-9
EINECS: 225-465-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR