3-Chlor-2-benzo[b]thiophencarbonsäure

Lieferant: MENAI

E2686.10
MENAE2686.10EA 0 EUR
MENAE2686.10
3-Chlor-2-benzo[b]thiophencarbonsäure
3-Chlor-2-benzo[b]thiophencarbonsäure
Formel: C₉H₅ClO₂S
Molecular Weight: 212,65 g/mol
MDL: MFCD00014579
CAS-Nummer: 21211-22-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR