5-Iodisatin

Lieferant: MENAI

E2692.5 E2692.25
MENAE2692.5EA 0 EUR
MENAE2692.5 MENAE2692.25
5-Iodisatin
5-Iodisatin
Formel: C₈H₄INO₂
MDL: MFCD00016899
CAS-Nummer: 20780-76-1
EINECS: 244-035-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR