2-Brom-3',5'-bis(trifluormethyl)acetophenon

Lieferant: MENAI

Synonyme: 1-(3,5-Bis(trifluoromethyl)phenyl)-2-bromoethanone, 3,5-Bis(trifluoromethyl)phenacyl bromide

E2695.10
MENAE2695.10EA 0 EUR
MENAE2695.10
2-Brom-3',5'-bis(trifluormethyl)acetophenon
2-Brom-3',5'-bis(trifluormethyl)acetophenon
Formel: C₁₀H₅BrF₆O
Molecular Weight: 335,0405 g/mol
MDL: MFCD00792434
CAS-Nummer: 131805-94-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR