Tris[3,5-bis(trifluormethyl)phenyl]phosphin

Lieferant: MENAI

Synonyme: Tris(3,5-bis(trifluoromethyl)phenyl)phosphine

E2908.5
MENAE2908.5EA 0 EUR
MENAE2908.5
Tris[3,5-bis(trifluormethyl)phenyl]phosphin
Tris[3,5-bis(trifluormethyl)phenyl]phosphin
Formel: C₂₄H₉F₁₈P
Molecular Weight: 670,28 g/mol
MDL: MFCD00042428
CAS-Nummer: 175136-62-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR