Methyl-3-chlor-2-benzo[b]thiophencarboxylat

Lieferant: MENAI

Synonyme: Methyl 3-chlorobenzo[b]thiophene-2-carboxylate

E3709.10
MENAE3709.10EA 0 EUR
MENAE3709.10
Methyl-3-chlor-2-benzo[b]thiophencarboxylat
Methyl-3-chlor-2-benzo[b]thiophencarboxylat
Formel: C₁₀H₇ClO₂S
Molecular Weight: 226.68 g/mol
MDL: MFCD00052503
CAS-Nummer: 21211-07-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR