2-Chlor-3'-nitroacetanilid

Lieferant: MENAI

E3715.50
MENAE3715.50EA 0 EUR
MENAE3715.50
2-Chlor-3'-nitroacetanilid
2-Chlor-3'-nitroacetanilid
Formel: C₈H₇ClN₂O₃
Molecular Weight: 214,61 g/mol
MDL: MFCD00024359
CAS-Nummer: 10147-71-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR