2-Brom-6-chlor-4-(trifluormethyl)anilin
Lieferant: MENAI
Synonyme:
4-Amino-3-brom-5-chlor-α,α,α-trifluortoluol, 4-Amino-3-brom-5-chlorbenzotrifluorid
E3866.5
MENAE3866.5EA
0
EUR
MENAE3866.5
2-Brom-6-chlor-4-(trifluormethyl)anilin
2-Brom-6-chlor-4-(trifluormethyl)anilin
Formel:
C₇H₄BrClF₃N Molecular Weight: 274,47 g/mol Siedepunkt: 71…72 °C (0,25 mmHg) Schmelzpunkt: 29…32 °C |
MDL:
MFCD00042180 CAS-Nummer: 109919-26-8 UN: 2811 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...