3-Bromphenylharnstoff

Lieferant: MENAI

Synonyme: 1-(3-Bromophenyl)urea

E3869.100
MENAE3869.100EA 0 EUR
MENAE3869.100
3-Bromphenylharnstoff
3-Bromphenylharnstoff
Formel: C₇H₇BrN₂O
Molecular Weight: 215.06 g/mol
MDL: MFCD00041317
CAS-Nummer: 2989-98-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR