2-Amino-4-(4-bromphenyl)thiazol

Lieferant: MENAI

E445.25
MENAE445.25EA 0 EUR
MENAE445.25
2-Amino-4-(4-bromphenyl)thiazol
2-Amino-4-(4-bromophenyl)thiazole
Formel: C₉H₇BrN₂S
Molecular Weight: 255,1343 g/mol
MDL: MFCD00051953
CAS-Nummer: 2103-94-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR