4-[(Trifluormethyl)thio]anilin

Lieferant: MENAI

Synonyme: 4-((Trifluoromethyl)thio)aniline

E906.5
MENAE906.5EA 0 EUR
MENAE906.5
4-[(Trifluormethyl)thio]anilin
4-[(Trifluormethyl)thio]anilin
Formel: C₇H₆F₃NS
Molecular Weight: 193,19 g/mol
MDL: MFCD00040926
CAS-Nummer: 372-16-7
EINECS: 206-744-4
UN: 2810
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR