p-Toluolsulfonylacetonitril

Lieferant: MENAI

Synonyme: [(4-Methylphenyl)sulphonyl]acetonitrile

ES488.25
MENAES488.25EA 0 EUR
MENAES488.25
p-Toluolsulfonylacetonitril
p-Toluolsulfonylacetonitril
Formel: C₉H₉NO₂S
Molecular Weight: 195,24 g/mol
MDL: MFCD00039487
CAS-Nummer: 5697-44-9
EINECS: 227-173-7
UN: 3439
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR