2-Chlor-5-nitrozimtsäure

Lieferant: MENAI
Achtung

Synonyme: 2-Chlor-5-nitrocinnamsäure, 3-(2-Chlor-5-nitrophenyl)propensäure

IP30.50
MENAIP30.50EA 0 EUR
MENAIP30.50
2-Chlor-5-nitrozimtsäure
2-Chlor-5-nitrozimtsäure
Formel: C₉H₆ClNO₄
Molecular Weight: 227.6 g/mol
Schmelzpunkt: 223…234 °C
MDL: MFCD00063312
CAS-Nummer: 36015-19-7
UN: 3077
ADR: 9 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR