2,4-Dichlorbenzamid

Lieferant: MENAI

Synonyme: 2,4-Dichlorbenzoesäureamid

IP34.100
MENAIP34.100EA 0 EUR
MENAIP34.100
2,4-Dichlorbenzamid
2,4-Dichlorbenzamid
Formel: C₇H₅Cl₂NO
Molecular Weight: 190.03 g/mol
Schmelzpunkt: 193…194 °C
MDL: MFCD00007974
CAS-Nummer: 2447-79-2
EINECS: 219-505-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR