2-Amino-3-brom-5-methylpyridin

Lieferant: MENAI
Achtung

LEH1.100
MENALEH1.100EA 0 EUR
MENALEH1.100
2-Amino-3-brom-5-methylpyridin
2-Amino-3-brom-5-methylpyridin
Formel: C₆H₇BrN₂
MDL: MFCD00068231
CAS-Nummer: 17282-00-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR