4-(Trifluormethoxy)benzolsulfonylchlorid
Lieferant: MENAI
Synonyme:
4-(Trifluormethoxy)benzolsulfonsäurechlorid, 4-(Trifluormethoxy)benzolsulfochlorid, 4-(Trifluormethoxy)benzol-1-sulfonylchlorid
NC1241.10
MENANC1241.10EA
0
EUR
MENANC1241.10
4-(Trifluormethoxy)benzolsulfonylchlorid
4-(Trifluormethoxy)benzolsulfonylchlorid
Formel:
C₇H₄ClF₃O₃S Molecular Weight: 260,62 g/mol Siedepunkt: 120…122 °C (15 mmHg) Dichte: 1,536 g/cm³ (20 °C) |
MDL:
MFCD00042408 CAS-Nummer: 94108-56-2 EINECS: 302-397-9 UN: 3265 ADR: 8,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...