(4-Fluorphenoxy)essigsäurehydrazid

Lieferant: MENAI

Synonyme: (4-Fluorphenoxy)acethydrazid, (4-Fluorphenoxy)acetohydrazid

NC127.25
MENANC127.25EA 0 EUR
MENANC127.25
(4-Fluorphenoxy)essigsäurehydrazid
(4-Fluorphenoxy)essigsäurehydrazid
Formel: (C₆H₄F)OCH₂CONHNH₂
Molecular Weight: 184,17 g/mol
MDL: MFCD00082651
CAS-Nummer: 1737-62-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR