5-Propyl-1,3-cyclohexandion

Lieferant: MENAI

NC1345.10
MENANC1345.10EA 0 EUR
MENANC1345.10
5-Propyl-1,3-cyclohexandion
5-Propyl-1,3-cyclohexanedione
Formel: C₉H₁₄O₂
MDL: MFCD00129456
CAS-Nummer: 57641-89-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR