trans-2-Pentensäure
Lieferant: MENAI
|
Gefahr
|
Synonyme:
(E)-Pent-2-enoic acid, 2-Pentenoic acid
NC1360.50
MENANC1360.50EA
0
EUR
MENANC1360.50
trans-2-Pentensäure
trans-2-Pentenoic acid
Formel:
C₅H₈O₂ Molecular Weight: 100,12 g/mol Siedepunkt: 205 °C (1013 hPa) Dichte: 0,9903 g/cm³ (20 °C) |
MDL:
MFCD00002704 CAS-Nummer: 13991-37-2 EINECS: 237-791-9 UN: 3265 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...