5-Isobutyl-1,3-cyclohexandion

Lieferant: MENAI

NC1414.10
MENANC1414.10EA 0 EUR
MENANC1414.10
5-Isobutyl-1,3-cyclohexandion
5-Isobutyl-1,3-cyclohexandion
Formel: C₁₀H₁₆O₂
MDL: MFCD00129458
CAS-Nummer: 57641-95-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR