4,5-Dichlorimidazol

Lieferant: MENAI

Synonyme: 4,5-Dichlor-1H-imidazol

NC2062.50
MENANC2062.50EA 0 EUR
MENANC2062.50
4,5-Dichlorimidazol
4,5-Dichloroimidazole
Formel: C₃H₂Cl₂N₂
Molecular Weight: 136,97 g/mol
MDL: MFCD00005195
CAS-Nummer: 15965-30-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR