2-(p-Toluolsulfonyl)acetophenon

Lieferant: MENAI

Synonyme: 1-Phenyl-2-tosylethanone

NC2279.25
MENANC2279.25EA 0 EUR
MENANC2279.25
2-(p-Toluolsulfonyl)acetophenon
2-(p-Toluolsulfonyl)acetophenon
Formel: C₁₅H₁₄O₃S
Molecular Weight: 274,340 g/mol
MDL: MFCD00026002
CAS-Nummer: 31378-03-7
EINECS: 250-605-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR