2-Nitrobenzolsulfonamid

Lieferant: MENAI

Synonyme: 2-Nitrobenzolsulfonsäureamid

NC2430.50
MENANC2430.50EA 0 EUR
MENANC2430.50
2-Nitrobenzolsulfonamid
2-Nitrobenzolsulfonamid
Formel: C₆H₆N₂O₄S
Molecular Weight: 202,19 g/mol
Schmelzpunkt: 195…196,5 °C
MDL: MFCD00009807
CAS-Nummer: 5455-59-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR