(3,4-Dimethoxyphenyl)essigsäurehydrazid

Lieferant: MENAI

NC354.50
MENANC354.50EA 0 EUR
MENANC354.50
(3,4-Dimethoxyphenyl)essigsäurehydrazid
(3,4-Dimethoxyphenyl)essigsäurehydrazid
Formel: C₁₀H₁₄N₂O₃
Molecular Weight: 210,23 g/mol
MDL: MFCD00051701
CAS-Nummer: 60075-23-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR