4-Chlorphenylsulfonylaceton

Lieferant: MENAI

Synonyme: 1-(4-Chlorphenylsulfonyl)aceton

NC884.25 NC884.100
MENANC884.25EA 0 EUR
MENANC884.25 MENANC884.100
4-Chlorphenylsulfonylaceton
4-Chlorphenylsulfonylaceton
Formel: C₉H₉ClO₃S
Molecular Weight: 232.69 g/mol
MDL: MFCD00018663
CAS-Nummer: 5000-48-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR