4,7-Dichlorisatin
Lieferant: MENAI
|
Achtung
|
Synonyme:
4,7-Dichlor-2,3-indolindion, 4,7-Dichlor-1H-indol-2,3-dion, 4,7-Dichlor-2,3-dioxoindolin, 4,7-Dichlorindolin-2,3-dion
SR462.10
MENASR462.10EA
0
EUR
MENASR462.10
4,7-Dichlorisatin
4,7-Dichlorisatin
Formel:
C₈H₃Cl₂NO₂ Molecular Weight: 216,02 g/mol |
MDL:
MFCD00047214 CAS-Nummer: 18711-13-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...