2,5-Dichlorbenzhydrazid

Lieferant: MENAI

Synonyme: 2,5-Dichlorobenzoic hydrazide

SR465.10
MENASR465.10EA 0 EUR
MENASR465.10
2,5-Dichlorbenzhydrazid
2,5-Dichlorbenzhydrazid
Formel: C₇H₆Cl₂N₂O
Molecular Weight: 205,04 g/mol
MDL: MFCD00014754
CAS-Nummer: 67487-35-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR