1-(2,5-Dichlorphenyl)-2-thioharnstoff

Lieferant: MENAI

Synonyme: N-(2,5-Dichlorphenyl)thioharnstoff, 2,5-Dichlorphenylthioharnstoff

SR470.50
MENASR470.50EA 0 EUR
MENASR470.50
1-(2,5-Dichlorphenyl)-2-thioharnstoff
1-(2,5-Dichlorphenyl)-2-thioharnstoff
Formel: (C₆H₃Cl₂)NC(S)NH₂
Molecular Weight: 221,11 g/mol
Schmelzpunkt: 320 °C
MDL: MFCD00041154
CAS-Nummer: 4949-85-3
UN: 2811
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR