Tris(3-fluorphenyl)phosphin

Lieferant: MENAI

SR471.5
MENASR471.5EA 0 EUR
MENASR471.5
Tris(3-fluorphenyl)phosphin
Tris(3-fluorphenyl)phosphin
Formel: C₁₈H₁₂F₃P
Molecular Weight: 316.26 g/mol
MDL: MFCD00015722
CAS-Nummer: 23039-94-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR