2-Aminophenyl-2-pyridylketon

Lieferant: MENAI

SR472.1 SR472.2 SR472.5 SR472.10
MENASR472.1EA 0 EUR
MENASR472.1 MENASR472.2 MENASR472.5 MENASR472.10
2-Aminophenyl-2-pyridylketon
2-Aminophenyl-2-pyridylketon
Formel: C₁₂H₁₀N₂O
MDL: MFCD03839744
CAS-Nummer: 42471-56-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR