3-Chlor-2-benzo[b]thiophencarbonylchlorid

Lieferant: MENAI

Synonyme: 3-Chlorobenzo[b]thiophene-2-carbonyl chloride

SR482.25
MENASR482.25EA 0 EUR
MENASR482.25
3-Chlor-2-benzo[b]thiophencarbonylchlorid
3-Chlor-2-benzo[b]thiophencarbonylchlorid
Formel: C₉H₄Cl₂OS
Molecular Weight: 231,10 g/mol
MDL: MFCD00053069
CAS-Nummer: 21815-91-8
UN: 3261
ADR: 8,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR