Ethyl-2-amino-4-methylthiazol-5-carboxylat

Lieferant: MENAI

Synonyme: Ethyl 2-amino-4-methylthiazole-5-carboxylate

ST1157.100
MENAST1157.100EA 0 EUR
MENAST1157.100
Ethyl-2-amino-4-methylthiazol-5-carboxylat
Ethyl-2-amino-4-methylthiazol-5-carboxylat
Formel: C₇H₁₀N₂O₂S
Molecular Weight: 186,22 g/mol
MDL: MFCD00123414
CAS-Nummer: 7210-76-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR