1-Methylisatin
Lieferant: MENAI
Synonyme:
1-Methylindolin-2,3-dion, 1-Methyl-1H-indol-2,3-dion, N-Methyl-2,3-dioxoindolin, N-Methyl-2,3-dihydroindol-2,3-dion, N-Methyl-1H-indol-2,3-dion, N-Methylindolin-2,3-dion, 1-Methyl-2,3-indolindion, N-Methylisatin, 1-Methyl-2,3-dioxoindolin, N-Methyl-2,3-indolindion, 1-Methyl-2,3-dihydroindol-2,3-dion
ST1916.25
MENAST1916.25EA
0
EUR
MENAST1916.25
1-Methylisatin
1-Methylisatin
Formel:
C₉H₇NO₂ Molecular Weight: 161,16 g/mol Schmelzpunkt: 139 °C Dichte: 1,4 g/cm³ (20 °C) |
MDL:
MFCD00005812 CAS-Nummer: 2058-74-4 EINECS: 218-164-9 UN: 2811 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...