2-Brom-2-methylpropiophenon

Lieferant: MENAI

ST2332.25
MENAST2332.25EA 0 EUR
MENAST2332.25
2-Brom-2-methylpropiophenon
2-Brom-2-methylpropiophenon
Formel: C₁₀H₁₁BrO
MDL: MFCD00000124
CAS-Nummer: 10409-54-8
EINECS: 233-879-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR