5-Methyl-1,3-cyclohexandion

Lieferant: MENAI
Achtung

ST254.50
MENAST254.50EA 0 EUR
MENAST254.50
5-Methyl-1,3-cyclohexandion
5-Methyl-1,3-cyclohexandion
Formel: C₇H₁₀O₂
Lagertemperatur: Raumtemperatur
MDL: MFCD00010379
CAS-Nummer: 4341-24-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR