3,4-Dimethyl-o-phenylendiamin
Lieferant: MENAI
Synonyme:
1,2-Diamino-3,4-dimethylbenzol, 3,4-Dimethylbenzol-1,2-diamin, 3,4-Dimethyl-1,2-phenylendiamin
WB132.5
WB132.10
MENAWB132.5EA
0
EUR
MENAWB132.5
MENAWB132.10
3,4-Dimethyl-o-phenylendiamin
3,4-Dimethyl-o-phenylendiamin
Formel:
C₈H₁₂N₂ Siedepunkt: 287 °C (1013 hPa) Schmelzpunkt: 89 °C |
MDL:
MFCD00051709 CAS-Nummer: 41927-01-9 UN: 2811 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...