2-(4-Chlorphenylthio)benzaldehyd

Lieferant: MENAI

WB176.100 WB176.50
MENAWB176.100EA 0 EUR
MENAWB176.100 MENAWB176.50
2-(4-Chlorphenylthio)benzaldehyd
2-(4-Chlorophenylthio)benzaldehyde
Formel: C₁₃H₉ClOS
MDL: MFCD00051766
CAS-Nummer: 107572-07-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR