Hygostar Astro Hood Made of Micromesh, Nylon
Lieferant: MENSCH FRANZ
132500
MENS132500EA 245 EUR
MENS132500
Hygostar Astro Hood Made of Micromesh, Nylon
Einweg-Kleidung
Hygostar Astro Hood made of high-quality breathable nylon, micromesh.
  • High-quality, soft nylon
  • Breathable, fine micromesh for a high level of comfort
  • Latex-free, very soft and light texture
  • Prevents skin irritation and pressure
  • Suitable for contact with food industry and slaughterhouses
  • Meets the highest standards
The special feature of these astro hoods lies in the material: Micromesh made of nylon is particularly breathable, soft and elastic. The micromesh allows the head and skin to breathe. This means that the astro hoods are always comfortable and pleasantly light, even when worn for long periods. In addition, the face cutout is so flexible that you can wear the astro hood in three ways: either just the head covered with a bare face, or pulled over the mouth, or pulled over the mouth and nose at the same time so that only the eye area is exposed. The Micromesh astro hoods are therefore ideal for hygiene areas in the food industry and slaughterhouses.
Standards: CE PSA VO (EU) 2016-425, CAT 1 - Personal protective equipment category 1, food-safe.