Surgical Mask, PP
Lieferant: MENSCH FRANZ
29197
MENS29197EA 124 EUR
MENS29197
Surgical Mask, PP
Masken OP-Masken
Face mask with elastic bands made from non woven PP, 3-ply (inner and outer layer) and a meltblown fabric filter media.
  • Excellent filter performance >98% BFE
  • Non sterile, lint free, skin-friendly
  • Integrated nose clip, anatomically adjustable
  • Single use
  • Suitable for medical use
No ear pressure even when worn for long periods.
Standards: EN 14683 Typ II, food safe.