Kaliumpermanganat 99,0-100,5 Manganometry, EMSURE® Reag. Ph. Eur., ACS analysis_for, Supelco®

Lieferant: Merck
Gefahr

Synonyme: Permanganic acid potassium salt

MQ300

1.05082.0250 1.05082.1000 1.05082.9050 1.05082.0250_N 1.05082.1000_N
1.05082.0250EA 48 EUR
1.05082.0250 1.05082.1000 1.05082.9050 MERC1.05082.0250_N MERC1.05082.1000_N
Kaliumpermanganat 99,0-100,5 Manganometry, EMSURE® Reag. Ph. Eur., ACS analysis_for, Supelco®
Potassium permanganate

For full details of specification and hazards please see www.vwr.com or the VWR Chemicals catalogues

Formel: KMnO₄
Molecular Weight: 158.03 g/mol
Schmelzpunkt: > 240 °C
Dichte: 2.7 g/cm³ (20 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00011364
CAS-Nummer: 7722-64-7
EINECS: 231-760-3
UN: 1490
ADR: 5.1 II
Merck Index: 12,07824

Bestellen

Spezifikation Testergebnisse

Assay (manganometric) 99.0 - 100.5 %
Identity Passes test
Appearance of solution Passes test
In water insoluble matter ≤ 0.2 %
Chloride (Cl) ≤ 0.02 %
Chloride, Chlorate (as Cl) ≤ 0.005 %
Sulfate (SO₄) ≤ 0.01 %
Total nitrogen (N) ≤ 0.003 %
Cu (Copper) ≤ 0.001 %
Fe (Iron) ≤ 0.002 %
Pb (Lead) ≤ 0.002 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR