Iod 0.05 M in Pyridin, Novabiochem® oxidierendes Reagens, zur DNA/RNA Synthese

Lieferant: Merck
Gefahr


M-Clarity™ Qualitätsstufe = MQ200

8.57008.1000 8.57008.2500
MERC8.57008.1000EA 0 EUR
MERC8.57008.1000 MERC8.57008.2500
Iod 0.05 M in Pyridin, Novabiochem® oxidierendes Reagens, zur DNA/RNA Synthese
Iod
Formel: I₂
Dichte: 0,99 g/cm³
Flammpunkt: 20,0 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00011355
CAS-Nummer: 7553-56-2
UN: 1282
ADR: 3,II

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR