Chlorpyrifos-methyl, Supelco®
Lieferant: Merck
|
Achtung
|
Synonyme:
O,O-Dimethyl-O-3,5,6-trichlor-2-pyridylphosphorothionat, O,O-Dimethyl-O-3,5,6-trichlor-2-pyridylthiophosphat
MET418B
MET418BEA
179
EUR
MET418B
Chlorpyrifos-methyl, Supelco®
Chlorpyrifos-methyl
Formel:
C₇H₇Cl₃NO₃PS Molecular Weight: 322,54 g/mol Schmelzpunkt: 45,5…46.5 °C |
CAS-Nummer:
5598-13-0 EINECS: 227-011-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...